Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GARNL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286779.1 | NP_001273708.1 | 5, 6 | 711 | ||
NM_032293.4 | NP_115669.3 | 4, 5 | 848 | |||
NR_104591.1 | 4, 5 | 848 | ||||
XM_005252267.3 | XP_005252324.1 | 6, 7 | 703 | |||
XM_005252268.2 | XP_005252325.1 | 6, 7 | 765 | |||
XM_011519086.2 | XP_011517388.1 | 7, 8 | 775 | |||
XM_011519087.2 | XP_011517389.1 | 7, 8 | 758 | |||
XM_011519089.2 | XP_011517391.1 | 6, 7 | 661 | |||
XM_011519090.2 | XP_011517392.1 | 5, 6 | 544 | |||
XM_017015202.1 | XP_016870691.1 | 5, 6 | 741 | |||
GenBank mRNA | AK095679.1 | - | 4, 5 | 743 | ||
AK293732.1 | - | 4, 5 | 746 | |||
AK294970.1 | - | 4, 5 | 350 | |||
AK296246.1 | - | 5, 6 | 711 | |||
AL136573.1 | - | 4, 5 | 402 | |||
BC034983.2 | - | 4, 5 | 458 | 4821143 |