Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM246 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303107.1 | NP_001290036.1 | 3 | 1759 | ||
NM_001303108.1 | NP_001290037.1 | 2 | 1634 | |||
NM_032342.2 | NP_115718.1 | 2 | 1704 | |||
XM_017015206.1 | XP_016870695.1 | 2 | 1894 | |||
GenBank mRNA | BC006115.1 | - | 2 | 1676 | 3503592 | |
BC033550.1 | - | 4 | 2502 | 4822472 | ||
BC113967.1 | - | 1 | 881 |