Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032364.5 | NP_115740.5 | 2 | 342 | ||
GenBank mRNA | AK057354.1 | - | 2 | 473 | ||
AK291222.1 | - | 2 | 577 | |||
AY188447.1 | - | 2 | 259 | |||
BC117146.1 | - | 2 | 213 | 40125697 | ||
BC117148.1 | - | 2 | 213 | 40125699 | ||
DA127901.1 | - | 3 | 422 | |||
EF560741.1 | - | 2 | 264 |