Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032364.5 | NP_115740.5 | 7 | 2917 | ||
GenBank mRNA | AK055945.1 | - | 6 | 1955 | ||
AK057354.1 | - | 3 | 1672 | |||
AK123025.1 | - | 6 | 2497 | |||
AY188447.1 | - | 7 | 2834 | |||
BC080655.1 | - | 6 | 1869 |