Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKT1S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098632.2 | NP_001092102.1 | 5 | 1428 | ||
NM_001098633.3 | NP_001092103.1 | 5 | 1216 | |||
NM_001278159.1 | NP_001265088.1 | 5 | 1441 | |||
NM_001278160.1 | NP_001265089.1 | 5 | 1252 | |||
NM_032375.5 | NP_115751.3 | 5 | 1850 | |||
GenBank mRNA | AK055511.1 | - | 5 | 1441 | ||
AK092610.1 | - | 4 | 1785 | |||
AK123687.1 | - | 5 | 1815 | |||
BC000031.2 | - | 3 | 645 | |||
BC007416.2 | - | 5 | 1736 | 2988648 | ||
BC015562.1 | - | 6 | 1020 | |||
BC051844.2 | - | 5 | 1320 | 6164197 |