Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032494.2 | NP_115883.2 | 8, 9 | 1005 | ||
XM_011511998.1 | XP_011510300.1 | 8, 9 | 950 | |||
XM_017005107.1 | XP_016860596.1 | 8, 9 | 958 | |||
GenBank mRNA | AF334161.1 | - | 8, 9 | 991 | ||
BC032001.1 | - | 8, 9 | 942 | 4749794 | ||
CV574571.1 | - | 6, 7 | 551 |