Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321630.1 | NP_001308559.1 | 2 | 1119 | ||
NM_032899.6 | NP_116288.2 | 3 | 1543 | |||
NM_207006.2 | NP_996889.1 | 3 | 2628 | |||
XM_011517338.1 | XP_011515640.1 | 2 | 916 | |||
GenBank mRNA | AF497803.1 | - | 2 | 868 | ||
AK095301.1 | - | 3 | 2628 | |||
AK098407.1 | - | 2 | 217 | |||
BC052300.1 | - | 2 | 932 | 6421272 | ||
DQ280323.1 | - | 3 | 1516 |