Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288587.2 | NP_001275516.1 | 2 | 1018 | ||
NM_001321630.1 | NP_001308559.1 | 3 | 1186 | |||
NM_032899.6 | NP_116288.2 | 4 | 1610 | |||
NM_207006.2 | NP_996889.1 | 4 | 2695 | |||
XM_005251087.2 | XP_005251144.1 | 2 | 815 | |||
XM_011517338.1 | XP_011515640.1 | 3 | 983 | |||
GenBank mRNA | AF497803.1 | - | 3 | 935 | ||
AK095301.1 | - | 4 | 2695 | |||
AK098407.1 | - | 3 | 284 | |||
BC007828.2 | - | 2 | 689 | 3610547 | ||
BC052300.1 | - | 3 | 999 | 6421272 | ||
DQ280322.1 | - | 2 | 728 | |||
DQ280323.1 | - | 4 | 1583 |