Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301627.1 | NP_001288556.1 | 12 | 2408 | ||
NM_052917.3 | NP_443149.2 | 13 | 2718 | |||
XM_011510537.2 | XP_011508839.1 | 14 | 2911 | |||
XM_011510538.2 | XP_011508840.1 | 12 | 2323 | |||
XM_017003258.1 | XP_016858747.1 | 14 | 2765 | |||
XM_017003259.1 | XP_016858748.1 | 13 | 2490 | |||
XM_017003260.1 | XP_016858749.1 | 14 | 2881 | |||
XM_017003261.1 | XP_016858750.1 | 13 | 2577 | |||
XM_017003263.1 | XP_016858752.1 | 11 | 2125 | |||
GenBank mRNA | AB067505.1 | - | 11 | 2018 | ||
AK308615.1 | - | 2 | 894 | |||
BC101031.1 | - | 11 | 2136 | 40007710 | ||
BC101032.2 | - | 12 | 2304 | 40007711 | ||
BC101033.1 | - | 11 | 2136 | 40007712 | ||
BC101034.2 | - | 11 | 2136 | 40007713 |