Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301627.1 | NP_001288556.1 | 12 | 4123 | ||
NM_052917.3 | NP_443149.2 | 13 | 4433 | |||
XM_011510537.2 | XP_011508839.1 | 14 | 4626 | |||
XM_011510538.2 | XP_011508840.1 | 12 | 4038 | |||
XM_017003258.1 | XP_016858747.1 | 14 | 4480 | |||
XM_017003259.1 | XP_016858748.1 | 13 | 4205 | |||
XM_017003260.1 | XP_016858749.1 | 14 | 4596 | |||
XM_017003261.1 | XP_016858750.1 | 13 | 4292 | |||
XM_017003263.1 | XP_016858752.1 | 11 | 3840 | |||
GenBank mRNA | AB067505.1 | - | 11 | 3733 |