Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004616.2 | NP_004607.1 | 5 | 494 | ||
XM_006719583.3 | XP_006719646.1 | 5 | 448 | |||
XM_017019913.1 | XP_016875402.1 | 4 | 530 | |||
GenBank mRNA | AK223022.1 | - | 4 | 422 | ||
AK313112.1 | - | 4 | 477 | |||
BC005246.1 | - | 5 | 494 | 3934330 | ||
BC070168.1 | - | 9 | 868 | |||
M35252.1 | - | 4 | 452 |