Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPRY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305163.1 | NP_001292092.1 | 4 | 1402 | ||
NM_001305164.1 | NP_001292093.1 | 4 | 781 | |||
NM_133368.2 | NP_588609.1 | 4 | 803 | |||
XM_005256220.1 | XP_005256277.1 | 4 | 1110 | |||
XM_011523427.1 | XP_011521729.1 | 5 | 889 | |||
XM_011523428.1 | XP_011521730.1 | 5 | 1203 | |||
XM_011523430.1 | XP_011521732.1 | 5 | 830 | |||
XM_017023844.1 | XP_016879333.1 | 5 | 1576 | |||
GenBank mRNA | AB075852.1 | - | 4 | 723 | ||
AK027549.1 | - | 4 | 1353 | |||
AK172845.1 | - | 4 | 759 | |||
BC013173.2 | - | 4 | 758 | 4340287 | ||
DB083249.1 | - | 5 | 520 |