Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314033.1 | NP_001300962.1 | 3 | 967 | ||
NM_133444.2 | NP_597701.1 | 3 | 719 | |||
GenBank mRNA | AB075831.1 | - | 3 | 650 | ||
AK122649.1 | - | 3 | 643 | |||
BC013013.2 | - | 3 | 641 | 3531734 | ||
JQ313906.1 | - | 1 | 494 |