Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BATF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300807.1 | NP_001287736.1 | 2 | 1861 | ||
NM_001300808.1 | NP_001287737.1 | 2 | 1742 | |||
NM_138456.3 | NP_612465.3 | 3 | 1929 | |||
GenBank mRNA | AK092453.1 | - | 2 | 1739 | ||
AK096702.1 | - | 3 | 1929 | |||
BC012330.1 | - | 2 | 1719 | 4638025 | ||
HM134154.1 | - | 3 | 1929 |