Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DPH7 SIRNA Assays ›
Gene Symbol:
DPH7
Gene Name:
diphthamide biosynthesis 7
Gene Aliases:
C9orf112, RRT2, WDR85
Chromosome Location:
Chr.9: 137554899 - 137578935 on Build GRCh38
UniGene ID:
Hs.292570
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
149119
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DPH7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_138778.3
NP_620133.1
9
1807
XM_005266123.2
XP_005266180.1
10
2265
XM_005266126.3
XP_005266183.1
8
2038
XM_005266128.2
XP_005266185.1
10
1891
XM_005266129.2
XP_005266186.1
10
1894
XM_005266130.2
XP_005266187.1
8
1662
XM_011519188.1
XP_011517490.1
8
1730
XM_011519189.1
XP_011517491.1
9
1908
XM_011519191.2
XP_011517493.1
9
2963
XM_011519193.2
XP_011517495.1
9
3012
XM_011519199.2
XP_011517501.1
9
1753
XM_011519200.1
XP_011517502.1
9
1756
XM_017015289.1
XP_016870778.1
8
2897
XM_017015290.1
XP_016870779.1
8
2946
XM_017015291.1
XP_016870780.1
10
2100
XM_017015292.1
XP_016870781.1
9
2170
XM_017015293.1
XP_016870782.1
9
2127
XM_017015294.1
XP_016870783.1
11
2080
XM_017015295.1
XP_016870784.1
9
2167
XM_017015296.1
XP_016870785.1
9
2006
XM_017015297.1
XP_016870786.1
11
2077
XM_017015298.1
XP_016870787.1
10
2360
XM_017015299.1
XP_016870788.1
10
1617
XM_017015300.1
XP_016870789.1
10
2357
XM_017015301.1
XP_016870790.1
8
2030
XM_017015302.1
XP_016870791.1
8
3084
XM_017015303.1
XP_016870792.1
9
2265
XM_017015304.1
XP_016870793.1
8
1730
XM_017015305.1
XP_016870794.1
8
3061
XM_017015306.1
XP_016870795.1
9
2199
XM_017015307.1
XP_016870796.1
9
2196
XM_017015308.1
XP_016870797.1
7
2923
XM_017015309.1
XP_016870798.1
9
1923
GenBank mRNA
AJ420445.1
-
1
1166
AK075115.1
-
9
1808
AK094389.1
-
1
1808
BC003123.1
-
9
1746
3502107
BC017335.2
-
9
1766
4642600
BC040173.1
-
2
1269
4791980
Back To Top
More Information
Panther Classification:
Molecular Function -
methyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
peptidyl-diphthamide biosynthetic process from peptidyl-histidine
diphthine methylesterase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-77psw:80/100.66.77.247:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline