Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138983.2 | NP_620450.2 | 1 | 909 | ||
GenBank mRNA | AB209791.1 | - | 1 | 903 | ||
AL355743.1 | - | 1 | 53 | |||
AL834450.1 | - | 1 | 902 | |||
BC026989.1 | - | 1 | 786 | 4944525 | ||
BC033290.1 | - | 1 | 916 | 4940343 |