Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRFN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152447.3 | NP_689660.2 | 3 | 1866 | ||
XM_005267369.3 | XP_005267426.1 | 3 | 4743 | |||
XM_017021048.1 | XP_016876537.1 | 4 | 4682 | |||
XM_017021049.1 | XP_016876538.1 | 2 | 2568 | |||
XM_017021050.1 | XP_016876539.1 | 2 | 4567 | |||
GenBank mRNA | AK055365.1 | - | 3 | 1186 | ||
AK096627.1 | - | 3 | 1839 | |||
AK299632.1 | - | 3 | 1180 | |||
AL832129.1 | - | 5 | 3100 | |||
BC043165.1 | - | 3 | 1291 | 5286818 |