Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152527.4 | NP_689740.2 | 4 | 1389 | ||
XM_005246353.3 | XP_005246410.1 | 3 | 1245 | |||
XM_011510751.2 | XP_011509053.1 | 4 | 1160 | |||
XM_011510752.1 | XP_011509054.1 | 4 | 1389 | |||
XM_017003480.1 | XP_016858969.1 | 4 | 1083 | |||
GenBank mRNA | AK055356.1 | - | 2 | 646 | ||
AK123207.1 | - | 5 | 1146 | |||
AK292873.1 | - | 4 | 1121 | |||
BC034382.1 | - | 4 | 1297 | |||
BC065524.1 | - | 4 | 1293 | 6058847 | ||
BX648181.1 | - | 2 | 557 |