Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOSPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177475.1 | NP_001170946.1 | 14 | 2142 | ||
NM_152581.3 | NP_689794.1 | 15 | 2178 | |||
GenBank mRNA | AK057941.1 | - | 1 | 145 | ||
AL834345.1 | - | 7 | 1230 | |||
BC030641.1 | - | 15 | 2145 | 4821861 |