Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152653.3 | NP_689866.1 | 6 | 1127 | ||
XM_005265432.2 | XP_005265489.1 | 6 | 1211 | |||
GenBank mRNA | AK057886.1 | - | 6 | 1128 | ||
AK307075.1 | - | 5 | 924 | |||
BC022332.1 | - | 6 | 1104 | 4295679 | ||
CB851345.1 | - | 1 | 115 |