Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040272.5 | NP_001035362.3 | 24 | 4424 | ||
XM_011518063.2 | XP_011516365.1 | 26 | 4562 | |||
XM_011518064.2 | XP_011516366.1 | 25 | 4677 | |||
XM_011518067.1 | XP_011516369.1 | 16 | 3292 | |||
XM_011518068.2 | XP_011516370.1 | 14 | 3895 | |||
XM_011518070.2 | XP_011516372.1 | 14 | 3060 | |||
XM_017015310.1 | XP_016870799.1 | 25 | 11199 | |||
XM_017015311.1 | XP_016870800.1 | 25 | 4511 | |||
XM_017015312.1 | XP_016870801.1 | 24 | 4627 | |||
XM_017015313.1 | XP_016870802.1 | 25 | 4325 | |||
GenBank mRNA | AK092602.1 | - | 3 | 483 | ||
BC100788.2 | - | 3 | 473 | |||
BC100789.2 | - | 3 | 475 | |||
BC100790.2 | - | 3 | 472 | |||
BX640951.1 | - | 5 | 557 |