Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040272.5 | NP_001035362.3 | 29 | 6504 | ||
XM_011518063.2 | XP_011516365.1 | 31 | 6642 | |||
XM_011518064.2 | XP_011516366.1 | 30 | 6757 | |||
XM_011518067.1 | XP_011516369.1 | 21 | 5372 | |||
XM_011518068.2 | XP_011516370.1 | 19 | 5975 | |||
XM_011518070.2 | XP_011516372.1 | 19 | 5140 | |||
XM_017015310.1 | XP_016870799.1 | 30 | 13279 | |||
XM_017015311.1 | XP_016870800.1 | 30 | 6591 | |||
XM_017015312.1 | XP_016870801.1 | 29 | 6707 | |||
XM_017015313.1 | XP_016870802.1 | 30 | 6405 | |||
GenBank mRNA | AK092602.1 | - | 7 | 2355 | ||
AK123028.1 | - | 6 | 1989 | |||
AK128724.1 | - | 2 | 3322 | |||
BX640951.1 | - | 10 | 2637 |