Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CADM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167674.1 | NP_001161146.1 | 2, 3 | 845 | ||
NM_001167675.1 | NP_001161147.1 | 3, 4 | 872 | |||
NM_001256502.1 | NP_001243431.1 | 3, 4 | 392 | |||
NM_001256503.1 | NP_001243432.1 | 2, 3 | 242 | |||
NM_001256504.1 | NP_001243433.1 | 4, 5 | 419 | |||
NM_001256505.1 | NP_001243434.1 | 3, 4 | 392 | |||
NM_153184.3 | NP_694854.2 | 2, 3 | 225 | |||
XM_006713081.3 | XP_006713144.1 | 3, 4 | 735 | |||
XM_006713082.3 | XP_006713145.1 | 2, 3 | 838 | |||
XM_017006062.1 | XP_016861551.1 | 4, 5 | 777 | |||
XM_017006063.1 | XP_016861552.1 | 2, 3 | 425 | |||
GenBank mRNA | AB646739.1 | - | 3, 4 | 872 | ||
AB646740.1 | - | 3, 4 | 392 | |||
AB646741.1 | - | 3, 4 | 392 | |||
AB646742.1 | - | 4, 5 | 419 | |||
AB646743.1 | - | 2, 3 | 242 | |||
AF538973.1 | - | 2, 3 | 225 | |||
AK295885.1 | - | 3, 4 | 868 | |||
AL834270.1 | - | 2, 3 | 815 | |||
BC105999.1 | - | 3, 4 | 872 | 5259422 | ||
BC112136.1 | - | 2, 3 | 281 | 8327604 | ||
BC112138.1 | - | 2, 3 | 293 | 8327606 |