Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153252.4 | NP_694984.4 | 7 | 720 | ||
XM_005262113.3 | XP_005262170.1 | 7 | 722 | |||
XM_017029384.1 | XP_016884873.1 | 2 | 126 | |||
XM_017029385.1 | XP_016884874.1 | 7 | 722 | |||
GenBank mRNA | AY497046.1 | - | 7 | 720 | ||
AY497047.1 | - | 4 | 652 | |||
AY497048.1 | - | 7 | 720 | |||
AY497049.1 | - | 6 | 690 | |||
AY497050.1 | - | 6 | 569 | |||
AY497052.1 | - | 8 | 755 | |||
AY497053.1 | - | 6 | 569 | |||
AY497054.1 | - | 5 | 470 | |||
AY497055.1 | - | 8 | 755 | |||
AY497056.1 | - | 3 | 501 | |||
AY497057.1 | - | 3 | 501 | |||
AY497059.1 | - | 5 | 711 | |||
AY497060.1 | - | 2 | 402 | |||
BC111490.1 | - | 7 | 670 |