Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282745.1 | NP_001269674.1 | 7 | 789 | ||
NM_001282746.1 | NP_001269675.1 | 7 | 789 | |||
NM_173081.4 | NP_775104.2 | 7 | 789 | |||
XM_005252381.2 | XP_005252438.1 | 6 | 610 | |||
XM_011519350.2 | XP_011517652.1 | 7 | 772 | |||
XM_017015832.1 | XP_016871321.1 | 7 | 772 | |||
GenBank mRNA | AK057389.1 | - | 7 | 732 | ||
AK302985.1 | - | 7 | 776 | |||
AY436643.1 | - | 7 | 750 | |||
BC039312.1 | - | 7 | 782 | 5296302 |