Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173550.2 | NP_775821.2 | 4 | 530 | ||
XM_005251397.2 | XP_005251454.1 | 4 | 413 | |||
XM_005251398.3 | XP_005251455.1 | 4 | 652 | |||
XM_005251399.4 | XP_005251456.1 | 5 | 712 | |||
XM_011517788.2 | XP_011516090.1 | 4 | 642 | |||
XM_011517791.2 | XP_011516093.1 | 4 | 652 | |||
XM_011517796.2 | XP_011516098.1 | 2 | 59 | |||
XM_017014431.1 | XP_016869920.1 | 4 | 652 | |||
XM_017014432.1 | XP_016869921.1 | 4 | 652 | |||
XM_017014433.1 | XP_016869922.1 | 4 | 652 | |||
XM_017014434.1 | XP_016869923.1 | 4 | 652 | |||
XM_017014435.1 | XP_016869924.1 | 4 | 652 | |||
XM_017014436.1 | XP_016869925.1 | 4 | 652 | |||
XM_017014437.1 | XP_016869926.1 | 4 | 652 | |||
XM_017014438.1 | XP_016869927.1 | 4 | 651 | |||
XM_017014439.1 | XP_016869928.1 | 4 | 652 | |||
XM_017014441.1 | XP_016869930.1 | 4 | 651 | |||
XM_017014442.1 | XP_016869931.1 | 4 | 651 | |||
XM_017014443.1 | XP_016869932.1 | 2 | 54 | |||
GenBank mRNA | AK308284.1 | - | 4 | 413 | ||
AY422473.1 | - | 4 | 530 | |||
BC144214.1 | - | 3 | 307 | 9052734 | ||
BC150600.1 | - | 3 | 307 | 9056970 | ||
BX537933.1 | - | 4 | 391 | |||
CR936775.1 | - | 4 | 622 |