Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144877.2 | NP_001138349.1 | 2 | 206 | ||
NM_173690.4 | NP_775961.2 | 2 | 206 | |||
GenBank mRNA | AK093983.1 | - | 2 | 165 | ||
AK098024.1 | - | 2 | 143 | |||
BC104030.2 | - | 2 | 134 | 40025626 | ||
BC104031.1 | - | 2 | 134 | 40025627 | ||
BC104032.1 | - | 2 | 134 | 40025629 | ||
DB048788.1 | - | 2 | 206 |