Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173791.4 | NP_776152.1 | 1 | 418 | ||
XM_005269518.3 | XP_005269575.1 | 1 | 391 | |||
XM_011539265.1 | XP_011537567.1 | 1 | 391 | |||
XM_011539266.2 | XP_011537568.1 | 1 | 391 | |||
GenBank mRNA | AK091746.1 | - | 1 | 391 | ||
BC028375.1 | - | 1 | 380 | 4837939 |