Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174976.2 | NP_777636.2 | 3 | 1604 | ||
XM_006720119.3 | XP_006720182.1 | 3 | 1834 | |||
XM_011536661.2 | XP_011534963.1 | 3 | 5473 | |||
GenBank mRNA | AK095612.1 | - | 3 | 1320 | ||
BC035881.1 | - | 2 | 983 | |||
BC117676.2 | - | 2 | 1429 | 40069732 |