Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOSC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181503.2 | NP_852480.1 | 7, 8 | 663 | ||
XM_006719763.1 | XP_006719826.1 | 6, 7 | 794 | |||
GenBank mRNA | AF025438.1 | - | 6, 7 | 392 | ||
AF279901.1 | - | 8, 9 | 454 | |||
AK096810.1 | - | 5, 6 | 2303 | |||
AL050353.1 | - | 6, 7 | 1886 | |||
BC020773.1 | - | 7, 8 | 424 | 4808747 |