Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181721.2 | NP_859072.1 | 2 | 351 | ||
XM_017017575.1 | XP_016873064.1 | 2 | 656 | |||
XM_017017576.1 | XP_016873065.1 | 2 | 660 | |||
GenBank mRNA | AB094092.1 | - | 2 | 351 | ||
BC028191.1 | - | 2 | 350 | 5167039 | ||
BC038969.1 | - | 2 | 226 | 5164198 | ||
BC125040.1 | - | 2 | 346 | 40115641 |