Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243797.1 | NP_001230726.1 | 3 | 1004 | ||
NM_001243798.1 | NP_001230727.1 | 3 | 1009 | |||
NM_006022.3 | NP_006013.1 | 3 | 1346 | |||
NM_183422.3 | NP_904358.2 | 3 | 4328 | |||
GenBank mRNA | AB082525.1 | - | 3 | 4329 | ||
AJ222700.1 | - | 3 | 1241 | |||
AK027071.1 | - | 3 | 2883 | |||
BC000456.2 | - | 3 | 1262 | |||
BC009528.2 | - | 1 | 389 | |||
BC016867.1 | - | 3 | 1265 | |||
BC069207.1 | - | 3 | 1254 | |||
BC105277.1 | - | 3 | 1257 | |||
BC146664.1 | - | 3 | 1261 | |||
BC146679.1 | - | 3 | 1261 | |||
CR627459.1 | - | 4 | 2521 | |||
U35048.1 | - | 3 | 1232 |