Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMSAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043318.2 | NP_001036783.1 | 3 | 2098 | ||
NM_194278.3 | NP_919254.2 | 3 | 2477 | |||
XM_005268204.1 | XP_005268261.1 | 3 | 2477 | |||
XM_005268205.1 | XP_005268262.1 | 3 | 2033 | |||
XM_005268206.1 | XP_005268263.1 | 4 | 2250 | |||
GenBank mRNA | AK127785.1 | - | 3 | 2476 | ||
BC003121.1 | - | 2 | 1160 | |||
BC006451.1 | - | 2 | 932 | |||
BC006511.2 | - | 2 | 1728 | |||
BC009202.2 | - | 3 | 2098 | |||
BC015668.2 | - | 3 | 2111 | |||
BC025330.1 | - | 2 | 1728 |