Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMSAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043318.2 | NP_001036783.1 | 12 | 5277 | ||
NM_194278.3 | NP_919254.2 | 12 | 5656 | |||
XM_005268204.1 | XP_005268261.1 | 13 | 5360 | |||
XM_005268205.1 | XP_005268262.1 | 13 | 4916 | |||
XM_005268206.1 | XP_005268263.1 | 14 | 5133 | |||
GenBank mRNA | AK024978.1 | - | 1 | 158 | ||
AK095434.1 | - | 1 | 1120 | |||
AK308604.1 | - | 2 | 1209 | |||
BC036475.1 | - | 1 | 1231 | |||
BC047633.1 | - | 1 | 1226 |