Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TREML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198153.2 | NP_937796.1 | 3, 4 | 552 | ||
GenBank mRNA | AK090633.1 | - | 2, 3 | 144 | ||
AY358271.1 | - | 3, 4 | 555 | |||
BC101780.1 | - | 3, 4 | 540 | 8069286 | ||
BC105101.1 | - | 3, 4 | 540 | 8144104 | ||
BC143652.1 | - | 3, 4 | 540 | 9052166 |