Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198276.2 | NP_938017.2 | 1, 2 | 441 | ||
XM_011532693.2 | XP_011530995.1 | 1, 2 | 441 | |||
XM_011532694.2 | XP_011530996.1 | 1, 2 | 441 | |||
GenBank mRNA | AK091902.1 | - | 1, 2 | 441 | ||
BC047439.1 | - | 1, 2 | 313 | 5289095 | ||
DR760587.1 | - | 1, 2 | 478 |