Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198276.2 | NP_938017.2 | 4 | 1126 | ||
XM_017003575.1 | XP_016859064.1 | 5 | 1037 | |||
GenBank mRNA | AK091902.1 | - | 3 | 2075 | ||
AK124081.1 | - | 4 | 1986 | |||
BC047439.1 | - | 4 | 998 | 5289095 |