Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198469.3 | NP_940871.2 | 3 | 328 | ||
XM_005251877.4 | XP_005251934.1 | 3 | 324 | |||
XM_005251878.3 | XP_005251935.1 | 3 | 299 | |||
XM_011518457.2 | XP_011516759.1 | 3 | 324 | |||
XM_011518458.1 | XP_011516760.1 | 4 | 407 | |||
GenBank mRNA | AK128877.1 | - | 3 | 325 | ||
AK302081.1 | - | 3 | 302 | |||
BC133690.1 | - | 3 | 319 | 40119769 | ||
HY011325.1 | - | 3 | 329 |