Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198581.2 | NP_940983.2 | 9 | 1551 | ||
XM_006712519.2 | XP_006712582.1 | 9 | 1558 | |||
GenBank mRNA | AB111887.1 | - | 8 | 997 | ||
AK127776.1 | - | 6 | 620 | |||
AK131416.1 | - | 9 | 1537 | |||
BC155540.1 | - | 8 | 997 | |||
BC167154.1 | - | 8 | 997 |