Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201566.2 | NP_963860.1 | 2 | 627 | ||
GenBank mRNA | BC109286.1 | - | 2 | 315 | ||
BC131739.1 | - | 2 | 315 | |||
BC142618.1 | - | 2 | 627 | 40147405 |
Gene Symbol: JOSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX538323.1 | - | 751 |