Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEX3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174118.1 | NP_001167589.1 | 3 | 1998 | ||
NM_203304.3 | NP_976049.3 | 2 | 2185 | |||
XM_017026811.1 | XP_016882300.1 | 2 | 1985 | |||
GenBank mRNA | AB062435.1 | - | 1 | 199 | ||
AB107353.1 | - | 2 | 1778 | |||
AF458084.1 | - | 3 | 1470 | |||
BC113384.1 | - | 3 | 1469 | |||
BC113741.1 | - | 3 | 1469 |