Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IYD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164694.1 | NP_001158166.1 | 6 | 4750 | ||
NM_001164695.1 | NP_001158167.1 | 6 | 4699 | |||
NM_001318495.1 | NP_001305424.1 | 5 | 4570 | |||
NM_203395.2 | NP_981932.1 | 5 | 4638 | |||
NR_134655.1 | 6 | 4811 | ||||
GenBank mRNA | AY259176.1 | - | 5 | 4638 | ||
AY259177.1 | - | 6 | 4750 |