Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190458.1 | NP_001177387.1 | 2 | 192 | ||
NM_001193536.1 | NP_001180465.1 | 2 | 192 | |||
NM_203447.3 | NP_982272.2 | 3 | 407 | |||
XM_011518045.2 | XP_011516347.1 | 3 | 262 | |||
XM_011518046.2 | XP_011516348.1 | 3 | 305 | |||
XM_011518047.2 | XP_011516349.1 | 3 | 488 | |||
XM_011518048.2 | XP_011516350.1 | 2 | 227 | |||
XM_017015173.1 | XP_016870662.1 | 3 | 262 | |||
XM_017015174.1 | XP_016870663.1 | 3 | 342 | |||
GenBank mRNA | AK093572.1 | - | 2 | 190 | ||
AK310555.1 | - | 2 | 174 | |||
AL832270.1 | - | 2 | 156 | |||
BC130518.1 | - | 2 | 170 | 40146549 | ||
BC143929.1 | - | 2 | 170 | 9052447 | ||
DB147029.1 | - | 2 | 192 | |||
DR004867.1 | - | 3 | 494 | |||
HM991479.1 | - | 2 | 192 | |||
HM991480.1 | - | 2 | 188 | |||
KC736820.1 | - | 1 | 91 |