Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190458.1 | NP_001177387.1 | 46 | 6602 | ||
NM_001193536.1 | NP_001180465.1 | 47 | 6698 | |||
NM_203447.3 | NP_982272.2 | 48 | 6913 | |||
XM_011518045.2 | XP_011516347.1 | 47 | 6672 | |||
XM_011518046.2 | XP_011516348.1 | 48 | 6811 | |||
XM_011518047.2 | XP_011516349.1 | 48 | 6994 | |||
XM_011518048.2 | XP_011516350.1 | 47 | 6733 | |||
XM_011518049.2 | XP_011516351.1 | 35 | 5473 | |||
XM_017015173.1 | XP_016870662.1 | 48 | 6768 | |||
XM_017015174.1 | XP_016870663.1 | 48 | 6848 | |||
GenBank mRNA | AB191037.1 | - | 48 | 6913 | ||
AK024436.1 | - | 27 | 4045 | |||
AK074081.1 | - | 34 | 5032 | |||
AK090429.1 | - | 40 | 5901 | |||
AL583913.1 | - | 5 | 1120 | |||
AL832270.1 | - | 46 | 8754 | |||
AY117695.1 | - | 35 | 5590 | |||
BC019102.2 | - | 13 | 2322 | |||
BC112894.1 | - | 17 | 2778 | |||
EU668358.1 | - | 5 | 1120 |