Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC403312 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301851.1 | NP_001288780.1 | 2 | 2772 | ||
GenBank mRNA | BC036197.1 | - | 3 | 2743 | 5273873 |