Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM151B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205548.2 | NP_991111.2 | 6 | 1192 | ||
XM_006714564.3 | XP_006714627.2 | 6 | 1110 | |||
XM_011543234.2 | XP_011541536.1 | 6 | 1571 | |||
XM_011543235.2 | XP_011541537.1 | 5 | 946 | |||
XM_011543236.2 | XP_011541538.1 | 6 | 1322 | |||
XM_011543237.2 | XP_011541539.1 | 5 | 1493 | |||
XM_017009166.1 | XP_016864655.1 | 6 | 1509 | |||
XM_017009167.1 | XP_016864656.1 | 6 | 1519 | |||
XM_017009168.1 | XP_016864657.1 | 7 | 1238 | |||
XM_017009169.1 | XP_016864658.1 | 5 | 1018 | |||
XM_017009170.1 | XP_016864659.1 | 5 | 1443 | |||
GenBank mRNA | AY358256.1 | - | 6 | 1192 | ||
BC131563.1 | - | 6 | 1124 | 40109365 |