Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129998.2 | NP_001123470.1 | 3 | 408 | ||
NM_001319241.1 | NP_001306170.1 | 3 | 400 | |||
NM_001319242.1 | NP_001306171.1 | 3 | 400 | |||
NM_205852.3 | NP_995324.2 | 3 | 408 | |||
NR_135049.1 | 3 | 408 | ||||
XM_011520658.2 | XP_011518960.1 | 4 | 419 | |||
XM_011520663.2 | XP_011518965.1 | 4 | 499 | |||
XM_017019295.1 | XP_016874784.1 | 4 | 480 | |||
GenBank mRNA | AK128243.1 | - | 3 | 403 | ||
AY358810.1 | - | 3 | 408 | |||
BC036036.1 | - | 3 | 268 | |||
BC136805.1 | - | 3 | 348 | 9020795 | ||
BC136806.1 | - | 3 | 348 | 9020796 | ||
BG484230.1 | - | 674 | ||||
BM563861.1 | - | 267 | ||||
DQ368812.1 | - | 3 | 275 |