Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMIGD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319942.1 | NP_001306871.1 | 6 | 824 | ||
NM_206832.2 | NP_996663.1 | 7 | 928 | |||
XM_011524787.1 | XP_011523089.1 | 7 | 942 | |||
GenBank mRNA | AK172838.1 | - | 6 | 824 | ||
AK290266.1 | - | 7 | 901 | |||
AY358153.1 | - | 7 | 916 |