Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXW12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159929.1 | NP_001153401.1 | 5 | 603 | ||
NM_207102.2 | NP_996985.2 | 6 | 777 | |||
XM_017006224.1 | XP_016861713.1 | 5 | 790 | |||
GenBank mRNA | AY247969.1 | - | 6 | 777 | ||
BC101303.1 | - | 5 | 950 | 40024696 | ||
BC101305.1 | - | 5 | 547 | 40024702 | ||
BC101306.1 | - | 3 | 285 |