Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BICDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207311.2 | NP_997194.2 | 1, 2 | 462 | ||
XM_005253991.4 | XP_005254048.1 | 1, 2 | 487 | |||
XM_006719694.3 | XP_006719757.1 | 1, 2 | 489 | |||
XM_006719696.3 | XP_006719759.1 | 1, 2 | 488 | |||
XM_006719697.3 | XP_006719760.1 | 1, 2 | 489 | |||
XM_011538999.2 | XP_011537301.1 | 1, 2 | 491 | |||
XM_011539001.2 | XP_011537303.1 | 1, 2 | 500 | |||
GenBank mRNA | AK129960.1 | - | 2, 3 | 405 | ||
U88834.1 | - | 1, 2 | 427 |